β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
N7004-10G N7004-1G N7004-5G N7004-250MG N7004-25G
SIALN7004-10GEA 660 EUR
SIALN7004-10G SIALN7004-1G SIALN7004-5G SIALN7004-250MG SIALN7004-25G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
β-Nicotinamide adenine dinucleotide (NAD, oxidized form) hydrate
Formula: C₂₁H₂₇N₇O₁₄P₂·H₂O
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4