3,3',5,5'-Tetramethylbenzidine dihydrochloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Dihydrochloride TMB, TMB-HCl, TMB 2HCl, TMB dihydrochloride
M-Clarity™ quality level = MQ200
T3405-100TAB T3405-50TAB T3405-5TAB
SIALT3405-100TABEA 550 EUR
SIALT3405-100TAB SIALT3405-50TAB SIALT3405-5TAB
3,3',5,5'-Tetramethylbenzidine dihydrochloride, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidine dihydrochloride
Formula: C₁₆H₂₀N₂·2HCl
MW: 313.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012961
CAS Number: 64285-73-0
EINECS: 264-769-6