1,2-Bis(dichlorophosphino)ethane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

261920-1G 261920-5G
SIAL261920-1GEA 71.6 EUR
SIAL261920-1G SIAL261920-5G
1,2-Bis(dichlorophosphino)ethane, Sigma-Aldrich®
1,2-Bis(dichlorophosphino)ethane
Formula: C₂H₄Cl₄P₂
MW: 231.813 g/mol
MDL Number: MFCD00009609
CAS Number: 28240-69-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR