Diphenylphosphinic chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Diphenylphosphinyl chloride

M-Clarity™ quality level = MQ100

230235-10G 230235-50G
SIAL230235-10GEA 71.1 EUR
SIAL230235-10G SIAL230235-50G
Diphenylphosphinic chloride, Sigma-Aldrich®
Diphenylphosphinic chloride
Formula: C₁₂H₁₀ClOP
MW: 236.63 g/mol
MDL Number: MFCD00002077
CAS Number: 1499-21-4
EINECS: 216-107-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR