Diphenylphosphinic acid 99%
Supplier: Thermo Fisher Scientific
148710050 148710250
ACRO148710050EA 47.6 EUR
ACRO148710050 ACRO148710250
Diphenylphosphinic acid 99%
Diphenylphosphinic acid
Formula: C₁₂H₁₁O₂P
MW: 218.19 g/mol
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00002132
CAS Number: 1707-03-5
EINECS: 216-948-5
Order Now
Specification Test Results
Appearance White Crystalline solid
Infrared spectrum Conforms
Melting point 193 to 196 °C
Titration with NaOH ≥98.5 %