Pluronic® L-64, Sigma-Aldrich®

Supplier: Merck

Synonyms: Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), PEG-PPG-PEG, Pluronic® L-64, Synperonic® L64

M-Clarity™ quality level = MQ200

435449-1L 435449-250ML
SIAL435449-1LEA 134 EUR
SIAL435449-1L SIAL435449-250ML
Pluronic® L-64, Sigma-Aldrich®
Poloxamer 184 (Pluronic® L-64, Synperonic® L64)

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MW: 2900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082049
CAS Number: 9003-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR