5-Bromo-2,4-dimethoxypyrimidine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
457418-1G
457418-5G
SIAL457418-1GEA
67.6
EUR
SIAL457418-1G
SIAL457418-5G
5-Bromo-2,4-dimethoxypyrimidine, Sigma-Aldrich®
5-Bromo-2,4-dimethoxypyrimidine
Formula:
C₆H₇BrN₂O₂ MW: 219.0400 g/mol |
MDL Number:
MFCD00038016 CAS Number: 56686-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...