N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide with 1% tert-butylchlorodimethylsilane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide, Silylating mixture VI, MTBSTFA + 1% TBDMSCl
M-Clarity™ quality level = MQ100
375934-5ML 375934-25ML 375934-10ML 375934-10X1ML
SIAL375934-5MLEA 143 EUR
SIAL375934-5ML SIAL375934-25ML SIAL375934-10ML SIAL375934-10X1ML
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide with 1% tert-butylchlorodimethylsilane, Sigma-Aldrich®
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide with 1% tert-butylchlorodimethylsilane
Formula: C₉H₁₈F₃NOSi
MW: 241.33 g/mol
MDL Number: MFCD00009671
CAS Number: 77377-52-7
UN: 1993
ADR: 3,III