Ammonium bismuth citrate ≥99.0% for microbiology, Millipore®
Supplier: Merck
Danger
Synonyms: Bismuth Ammonium Citrate, Citric acid ammonium bismuth salt
M-Clarity™ quality level = MQ200
09678-100G 09678-500G
SIAL09678-100GEA 117 EUR
SIAL09678-100G SIAL09678-500G
Ammonium bismuth citrate ≥99.0% for microbiology, Millipore®
Ammonium bismuth citrate
Base ingredient for culture media
Formula: C₆H₁₁BiNO₇
MW: 433.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036420
CAS Number: 31886-41-6
EINECS: 250-856-6