FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 95%
Supplier: Thermo Scientific
Danger
Synonyms: 3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-FITC, Fluorescein Isothiocyanate, 5-isothiocyanatofluorescein, Fluorescein Isothiocyanate, Isomer I
L09319.03 L09319.MF L09319.MD
L09319.03EA 291 EUR
L09319.03 L09319.MF L09319.MD
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 95%
FITC (Fluorescein 5-isothiocyanate, Fluorescein isothiocyanate isomer I)
Purity: 95% CAS No.: 3326-32-7 Molecular Formula: C21H11NO5S Formula Weight: 389.39 Storage Temperature: 2°C to 8°C Melting Point: 360° Beilstein Reference: 1407295 MDL No.: MFCD00005063
Formula: C₂₁H₁₁NO₅S
MW: 389.381 g/mol
Melting Pt: >360 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005063
CAS Number: 3326-32-7
EINECS: 222-042-0