Erythromycin, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIALE5389-5GEA 138 EUR
SIALE5389-5G SIALE5389-1G
Erythromycin, Sigma-Aldrich®
Erythromycin
Formula: C₃₇H₆₇NO₁₃
MW: 733,95 g/mol
CAS Number: 114-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR