N-Methyl-N-octanoyl-D-glucamine (MEGA-8), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
O3129-25G
O3129-5G
SIALO3129-25GEA
2000
EUR
SIALO3129-25G
SIALO3129-5G
N-Methyl-N-octanoyl-D-glucamine (MEGA-8), Sigma-Aldrich®
N-Methyl-N-octanoyl-D-glucamine (MEGA-8)
Formula:
C₁₅H₃₁NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00134152 CAS Number: 85316-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...