Methyl hydrogen glutarate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-Methoxy-5-oxopentanoic acid

M-Clarity™ quality level = MQ100

M47353-25G M47353-5G
SIALM47353-25GEA 169 EUR
SIALM47353-25G SIALM47353-5G
Methyl hydrogen glutarate, Sigma-Aldrich®
Methyl hydrogen glutarate
Formula: C₆H₁₀O₄
MW: 146.1412 g/mol
MDL Number: MFCD00004409
CAS Number: 1501-27-5
EINECS: 216-116-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR