Creatine (N-amidinosarcosine), Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(1-Methylguanidino)acetic acid, N-Amidinosarcosine, Creatine

M-Clarity™ quality level = MQ200

C0780-100G C0780-10MG C0780-500G C0780-50G
SIALC0780-100GEA 246 EUR
SIALC0780-100G SIALC0780-10MG SIALC0780-500G SIALC0780-50G
Creatine (N-amidinosarcosine), Sigma-Aldrich®
Creatine (N-Amidinosarcosine)
Formula: C₄H₉N₃O₂
MW: 131.13 g/mol
MDL Number: MFCD00004282
CAS Number: 57-00-1
EINECS: 200-306-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR