Ammonium bismuth citrate Bi 48-52%, (max. 2% H₂O)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Bismuth Ammonium Citrate, Citric acid ammonium bismuth salt
A17003.30 A17003.18 A17003.0B
A17003.30EA 203 EUR
A17003.30 A17003.18 A17003.0B
Ammonium bismuth citrate Bi 48-52%, (max. 2% H₂O)
Ammonium bismuth citrate
CAS No.: 25530-63-6 Molecular Formula: C6H10BiNO8 Formula Weight: 433.12 MDL No.: MFCD00036420
Formula: C₆H₁₁BiNO₇
MW: 433.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036420
CAS Number: 25530-63-6
EINECS: 247-078-4