N-Cbz-glycine(Z-Gly-OH), Sigma-Aldrich®

Supplier: Merck

Synonyms: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

M-Clarity™ quality level = MQ200

C7206-100G C7206-25G C7206-500G
SIALC7206-100GEA 157 EUR
SIALC7206-100G SIALC7206-25G SIALC7206-500G
N-Cbz-glycine(Z-Gly-OH), Sigma-Aldrich®
N-Cbz-Glycine (Z-Gly-OH)
Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
MDL Number: MFCD00002691
CAS Number: 1138-80-3
EINECS: 214-516-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR