3-Quinolineboronic acid 97%
Supplier: Thermo Scientific
Warning
Synonyms: Quinoline-3-boronic acid, Quinolin-3-ylboronic acid
367730010 367730050 367730250
ACRO367730010EA 86 EUR
ACRO367730010 ACRO367730050 ACRO367730250
3-Quinolineboronic acid 97%
3-Quinolineboronic acid
Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Melting Pt: 148…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02183527
CAS Number: 191162-39-7
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Light pink to orange to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %