8-Quinolineboronic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: Quinolin-8-ylboronic acid

M-Clarity™ quality level = MQ100

542865-1G 542865-250MG
SIAL542865-1GEA 101 EUR
SIAL542865-1G SIAL542865-250MG
8-Quinolineboronic acid, Sigma-Aldrich®
8-Quinolineboronic acid
Formula: C₉H₈BNO₂
MW: 172.98 g/mol
MDL Number: MFCD01114698
CAS Number: 86-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR