Puromycin dihydrochloride

Supplier: Molekula

MOLE43504617-25MGEA 340 EUR
MOLE43504617-25MG
Puromycin dihydrochloride
Puromycin dihydrochloride
Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544,44 g/mol
MDL Number: MFCD00012691
CAS Number: 58-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR