Adenosine-5'-diphosphoric acid (ADP), Sigma-Aldrich®

Supplier: Merck

Synonyms: Adenosine 5'-diphosphate, 5′-ADP, Adenosine 5′-diphosphate

01905-250MG-F 01905-1G-F
SIAL01905-250MG-FEA 94.3 EUR
SIAL01905-250MG-F SIAL01905-1G-F
Adenosine-5'-diphosphoric acid (ADP), Sigma-Aldrich®
Adenosine-5'-diphosphoric acid (ADP)
Formula: C₁₀H₁₅N₅O₁₀P₂
MW: 427.2 g/mol
MDL Number: MFCD00066473
CAS Number: 58-64-0
EINECS: 200-392-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR