Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5′-AMP-Na2 hydrate
M-Clarity™ quality level = MQ100
01930-25G 01930-5G
SIAL01930-25GEA 229 EUR
SIAL01930-25G SIAL01930-5G
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate, Sigma-Aldrich®
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Formula: C₁₀H₁₂N₅Na₂O₇P·H₂O
MW: 391.2 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00065023
CAS Number: 4578-31-8
EINECS: 224-961-2