Nystatin dihydrate, 4.400 U/mg

Supplier: Roth Carl

0241.1 0241.2
ROTH0241.1EA 48.4 EUR
ROTH0241.1 ROTH0241.2
Nystatin dihydrate, 4.400 U/mg
Nystatin dihydrate
Formula: C₄₇H₇₅NO₁₇·2H₂O
MDL Number: MFCD00036240
CAS Number: 1400-61-9
EINECS: 215-749-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR