2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2) 100 mmol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2'-Deoxycytidine-5'-triphosphoric acid disodium salt, 2′-Deoxycytidine-5′-Triphosphate Trisodium Salt, 2'-Deoxycytidine-5'-triphosphate disodium salt
D4913-25UMO D4913-.1MMO
SIALD4913-25UMOEA 176 EUR
SIALD4913-25UMO SIALD4913-.1MMO
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2) 100 mmol/l in aqueous solution, Sigma-Aldrich®
2'-Deoxycytidine-5'-triphosphoric acid disodium salt (dCTP-Na2)
Formula: C₉H₁₄N₃Na₂O₁₃P₃
MW: 511.12 g/mol
MDL Number: MFCD00084683
CAS Number: 102783-51-7
EINECS: 252-849-3