n-Dodecyl-β-D-maltoside, Sigma-Aldrich®
Supplier: Merck
Synonyms: DDM
M-Clarity™ quality level = MQ200
SIALD4641-1GEA 356 EUR
SIALD4641-1G SIALD4641-500MG SIALD4641-25G SIALD4641-5G
n-Dodecyl-β-D-maltoside, Sigma-Aldrich®
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
MW: 510,6 g/mol
MDL Number: MFCD00043012
CAS Number: 69227-93-6