n-Nonyl-β-D-maltoside, Sigma-Aldrich®

Supplier: Merck

Synonyms: NM

SIAL59965-1GEA 203 EUR
SIAL59965-1G
n-Nonyl-β-D-maltoside, Sigma-Aldrich®
n-Nonyl-β-D-maltoside
Formula: C₂₁H₄₀O₁₁
MW: 468,54 g/mol
MDL Number: MFCD08062400
CAS Number: 106402-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR