Octyl-β-D-thioglucopyranoside
Supplier: BIOMOL RESEARCH LABORATORIES
Synonyms: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside
50739-2 50739-3
BMOL50739-2EA 0 EUR
BMOL50739-2 BMOL50739-3
Octyl-β-D-thioglucopyranoside
Octyl β-D-thioglucopyranoside
Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012189
CAS Number: 85618-21-9