Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride), Sigma-Aldrich®
Supplier: Merck
Synonyms: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl
MQ300
B8385-100MG B8385-10MG B8385-1MG B8385-25MG B8385-5MG
SIALB8385-100MGEA 2070 EUR
SIALB8385-100MG SIALB8385-10MG SIALB8385-1MG SIALB8385-25MG SIALB8385-5MG
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride), Sigma-Aldrich®
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Formula: C₁₆H₂₅ClN₂O₄
MW: 308.4 · 36.5 g/mol
MDL Number: MFCD00058004
CAS Number: 65391-42-6