Methotrexate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
M-Clarity™ quality level = MQ200
M9929-100MG M9929-500MG
SIALM9929-100MGEA 456 EUR
SIALM9929-100MG SIALM9929-500MG
Methotrexate, Sigma-Aldrich®
Methotrexate
Formula: C₂₀H₂₂N₈O₅
MW: 454.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00150847
CAS Number: 59-05-2
EINECS: 200-413-8
UN: 2811
ADR: 6.1,III