Mitomycin C (from Streptomyces caespitosus), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIALM0440-25MGEA 726 EUR
SIALM0440-25MG SIALM0440-5MG
Mitomycin C (from Streptomyces caespitosus), Sigma-Aldrich®
Mitomycin C
Formula: C₁₅H₁₈N₄O₅
MW: 334,33 g/mol
MDL Number: MFCD00078109
CAS Number: 50-07-7
UN: 2811
ADR: 6.1,II