Mitomycin C (from Streptomyces caespitosus), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALM0503-5X2MGEA
524
EUR
SIALM0503-5X2MG
SIALM0503-2MG
SIALM0503-50X2MG
SIALM0503-10X2MG
Mitomycin C (from Streptomyces caespitosus), Sigma-Aldrich®
Mitomycin C
Formula:
C₁₅H₁₈N₄O₅ MW: 334,33 g/mol |
MDL Number:
MFCD00078109 CAS Number: 50-07-7 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...