3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate, TMB-HCl hydrate, Dihydrochloride TMB hydrate
M-Clarity™ quality level = MQ200
T8768-100MG T8768-1G T8768-5G
SIALT8768-100MGEA 27.2 EUR
SIALT8768-100MG SIALT8768-1G SIALT8768-5G
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Formula: C₁₆H₂₀N₂·2HCl·xH₂O
MW: 313.27 g/mol (anhy)
MDL Number: MFCD00150104
CAS Number: 207738-08-7
EINECS: 264-769-6