EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ400
SIALE0276-25GEA 66 EUR
SIALE0276-25G SIALE0276-100G SIALE0276-500G
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid), Sigma-Aldrich®
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid)
Formula: C₉H₂₀N₂O₄S
MW: 252,33 g/mol
MDL Number: MFCD00006160
CAS Number: 16052-06-5