EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid), Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ400

SIALE1894-100GEA 476 EUR
SIALE1894-100G SIALE1894-1KG SIALE1894-25G SIALE1894-250G
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid), Sigma-Aldrich®
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid)
Formula: C₉H₂₀N₂O₄S
MW: 252,33 g/mol
MDL Number: MFCD00006160
CAS Number: 16052-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR