ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Supplier: Biotium
10050.EA 229 EUR
10050.
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Formula: C₁₈H₂₄N₆O₆S₄
MW: 548,68 g/mol
MDL Number: MFCD00010404
CAS Number: 30931-67-0