1-Allyl-3-methylimidazolium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Allyl-3-methyl-3-imidazolium chloride
43961-50G-F 43961-5G-F
SIAL43961-50G-FEA 280 EUR
SIAL43961-50G-F SIAL43961-5G-F
1-Allyl-3-methylimidazolium chloride, Sigma-Aldrich®
1-Allyl-3-methylimidazolium chloride
Formula: C₇H₁₁ClN₂
MW: 158.63 g/mol
MDL Number: MFCD07784450
CAS Number: 65039-10-3