Sudan black B, high purity biological stain
Supplier: Thermo Scientific
Synonyms: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
190160250 190161000
ACRO190160250EA 67.8 EUR
ACRO190160250 ACRO190161000
Sudan black B, high purity biological stain
Sudan black B
Formula: C₂₉H₂₄N₆
MW: 456.55 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006919
CAS Number: 4197-25-5
EINECS: 224-087-1
Order Now
Specification Test Results
Appearance Dark brown to black Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies