4,4'-Dimethyl-2,2'-bipyridyl 99+%
Supplier: Thermo Scientific
Warning
Synonyms: 4,4'-Dimethyl-2,2'-bipyridine
373230010 373230050 373230250
ACRO373230010EA 53.6 EUR
ACRO373230010 ACRO373230050 ACRO373230250
4,4'-Dimethyl-2,2'-bipyridyl 99+%
4,4'-Dimethyl-2,2'-bipyridyl
Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00006441
CAS Number: 1134-35-6
EINECS: 214-483-2
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige Crystalline powder
Infrared spectrum Conforms
GC ≥99.0 %