4,4'-Dinonyl-2,2'-bipyridyl, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4,4′-Dinonyl-2,2′-dipyridyl, 4,4'-Dinonyl-2,2'-bipyridine, 4,4'-Di-n-nonyl-2,2'-bipyridine

482250-1G 482250-5G
SIAL482250-1GEA 67.5 EUR
SIAL482250-1G SIAL482250-5G
4,4'-Dinonyl-2,2'-bipyridyl, Sigma-Aldrich®
4,4'-Dinonyl-2,2'-bipyridyl
Formula: C₂₈H₄₄N₂
MW: 408.67 g/mol
MDL Number: MFCD00800915
CAS Number: 142646-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR