Containers with screw cap
Supplier: LP ITALIANA
102048 102548 105047 105048 206047 206048 105148 206348 105548 105248 105147
LPIT102048EA 125 EUR
LPIT102048 LPIT102548 LPIT105047 LPIT105048 LPIT206047 LPIT206048 LPIT105148 LPIT206348 LPIT105548 LPIT105248 LPIT105147
Containers with screw cap
Sample Containers General Purpose Containers
PP containers with screw cap.
  • Good chemical resistance
  • Good temperature resistance
  • Available non steril or packed under aseptic conditions
Ideal for collecting, storing and transporting samples.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904