VWR®, Burettes, Lateral Stopcock
Supplier: VWR Collection
612-4144EA 65.2 EUR
612-4144 612-4142 612-4143
VWR®, Burettes, Lateral Stopcock
Burettes
Borosilicate 3.3 glass, class AS.
  • Schellbach stripe, stopcock with glass key
  • High contrast blue enamel graduations
  • Batch certificate included
DIN EN ISO 385
Certifikace: DIN EN ISO 385