Trimethoxysilane 95%
Supplier: Thermo Scientific
Danger
392170050 392170250 392171000
ACRO392170050EA 53.5 EUR
ACRO392170050 ACRO392170250 ACRO392171000
Trimethoxysilane 95%
Trimethoxysilane
Formula: C₃H₁₀O₃Si
MW: 122.2 g/mol
Boiling Pt: 81…83 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.96 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008340
CAS Number: 2487-90-3
EINECS: 219-637-2
UN: 3384
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3570 to 1.3600 (20°C, 589 nm)
GC ≥94 %