DL-Cysteine 96%
Supplier: Thermo Fisher Scientific
H56126.06 H56126.14
H56126.06EA 56.9 EUR
H56126.06 H56126.14
DL-Cysteine 96%
DL-Cysteine
Formula: C₃H₇NO₂S
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 225 °C (dec.)
Density: 1.441 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004881
CAS Number: 3374-22-9
EINECS: 222-160-2
Merck Index: 12,02850