2-Bromo-N-methylaniline 95%
Supplier: Thermo Fisher Scientific
Warning
H55166.03 H55166.06
H55166.03EA 114 EUR
H55166.03 H55166.06
2-Bromo-N-methylaniline 95%
2-Bromo-N-methylaniline
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 107…109 °C (12 mmHg)
Density: 1.589
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD06798064
CAS Number: 6832-87-7