4-Bromo-N-methylaniline ≥97%
Supplier: Thermo Fisher Scientific
Warning
H55273.03 H55273.06
H55273.03EA 61.6 EUR
H55273.03 H55273.06
4-Bromo-N-methylaniline ≥97%
4-Bromo-N-methylaniline
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 259…260 °C
Density: 1.482
Storage Temperature: Ambient
MDL Number: MFCD03030391
CAS Number: 6911-87-1