2,7-Dibromonaphthalene ≥99%
Supplier: Thermo Scientific
Warning
H56097.03 H56097.06
H56097.03EA 127 EUR
H56097.03 H56097.06
2,7-Dibromonaphthalene ≥99%
2,7-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285.97 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00238580
CAS Number: 58556-75-5
UN: 3077
ADR: 9,III