L(+)-Lactic acid 88 - 92% ≥88-92% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
M-Clarity™ quality level = MQ200
27714-1L 27714-2.5L 27714-25KG 27714-500ML
SIAL27714-1LEA 203 EUR
SIAL27714-1L SIAL27714-2.5L SIAL27714-25KG SIAL27714-500ML
L(+)-Lactic acid 88 - 92% ≥88-92% in aqueous solution, Sigma-Aldrich®
L(+)-Lactic acid
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 235
Melting Pt: 53
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 13,05352