4-Bromosalicylaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Bromo-2-hydroxybenzaldehyde
H54850.03 H54850.06
H54850.03EA 201 EUR
H54850.03 H54850.06
4-Bromosalicylaldehyde ≥97%
4-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 50…54 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD06252606
CAS Number: 22532-62-3
UN: 3077
ADR: 9 III