Dexamethasone
Supplier: Cayman Chemical
Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
11015-500 11015-1 11015-5
CAYM11015-500EA 76 EUR
CAYM11015-500 CAYM11015-1 CAYM11015-5
Dexamethasone
Dexamethasone
Formula: C₂₂H₂₉FO₅
Melting Pt: 268…271 °C
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9