1,3-Dicyclohexylurea
Supplier: Cayman Chemical
Synonyms: N,N′-Dicyclohexylurea, N,N'-Dicyclohexylurea
10004971-10 10004971-25
CAYM10004971-10EA 27 EUR
CAYM10004971-10 CAYM10004971-25
1,3-Dicyclohexylurea
1,3-Dicyclohexylurea
Formula: C₁₃H₂₄N₂O
MW: 224.35 g/mol
MDL Number: MFCD00003829
CAS Number: 2387-23-7
EINECS: 219-213-7