Zirconium(IV) oxide ≥99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Zirconium dioxide, Zirconium Oxide
A13578.22 A13578.0E A13578.36
A13578.22EA 25.4 EUR
A13578.22 A13578.0E A13578.36
Zirconium(IV) oxide ≥99%
Zirconium (IV) oxide
Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: 4300
Melting Pt: 2680
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180